missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dihydroxy-1,4-dithiane, 96%

Manufacturer:  Alfa Aesar™ A1462030

 View more versions of this product

Catalog No. AAA1462030



Specifications

Specifications

2,5-Dihydroxy-1,4-dithiane
250g
C4H8O2S2
UN2811
p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
C1C(SCC(S1)O)O
152.226
152.24
~130°C (decomposition)
40018-26-6
MFCD00006659
104402
YUIOPHXTILULQC-UHFFFAOYSA-N
1,4-dithiane-2,5-diol
98330
96%

Chemical Identifiers

40018-26-6
152.226
YUIOPHXTILULQC-UHFFFAOYSA-N
98330
C1C(SCC(S1)O)O
C4H8O2S2
MFCD00006659
p-dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithiane, mercaptoacetaldehyde dimer, 4-dithiane-2,5-diol, 1,4 dithiane-2,5-diol, 2,5-dihydroxy-1,4-dithian, p-dthane-2,5-dol, pubchem20480, 1,4dithiane-2,5diol, acmc-1anme
1,4-dithiane-2,5-diol
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H317
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P310-P301+P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents