Learn More
2,5-Dihydroxy-1,4-benzoquinone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1144206
Description
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2,5-Dihydroxy-1,4-benzoquinone | |
∼220°C (sublimation) | |
MFCD00001598 | |
1939229 | |
Soluble in water. | |
OC1=CC(=O)C(O)=CC1=O | |
140.09 | |
140.1 |
615-94-1 | |
C6H4O4 | |
5 g | |
2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop | |
QFSYADJLNBHAKO-UHFFFAOYSA-N | |
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione | |
69213 | |
98% |
Chemical Identifiers
615-94-1 | |
140.09 | |
QFSYADJLNBHAKO-UHFFFAOYSA-N | |
69213 | |
OC1=CC(=O)C(O)=CC1=O |
C6H4O4 | |
MFCD00001598 | |
2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop | |
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 210-454-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only