Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-2,5-Dihydro-D-phenylglycine, 97%

$47.77 - $464.08

Chemical Identifiers

Chemical Name or Material (-)-2,5-Dihydro-D-phenylglycine
CAS 26774-88-9
Molecular Formula C8H11NO2
Formula Weight 153.18
MDL Number MFCD00137746
Beilstein 2091420
Synonym (R)-alpha-Amino-1-(1,4-cyclohexadienyl)acetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1378506

  SDS
alfa aesar™
L1378506
5g Each for $47.77

AAL1378514

  SDS
alfa aesar™
L1378514
25g Each for $179.31

AAL1378522

  SDS
alfa aesar™
L1378522
100g Each for $464.08
Description & Specifications

Specifications

Formula Weight 153.18
CAS 26774-88-9
Chemical Name or Material (-)-2,5-Dihydro-D-phenylglycine
Synonym (R)-alpha-Amino-1-(1,4-cyclohexadienyl)acetic acid
Assay 97%
Molecular Formula C8H11NO2
Beilstein 2091420
Quantity 5g
Melting Point 288° to 290°C (subl.)
Optical Rotation −160° (c=1 in 1N HCl)
MDL Number MFCD00137746