missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (-)-2,5-Dihydro-D-phenylglycine, 97%

Manufacturer:  Alfa Aesar™ L1378514

 View more versions of this product

Catalog No. AAL1378514



Specifications

Specifications

(-)-2,5-Dihydro-D-phenylglycine
C8H11NO2
2091420
JBJJTCGQCRGNOL-SSDOTTSWSA-N
(2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid
1268102
97%
25g
26774-88-9
MFCD00137746
r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine
C1C=CCC(=C1)C(C(=O)O)N
153.181
153.18
288°C to 290°C (sublimation)
−160° (c=1 in 1N HCl)

Chemical Identifiers

26774-88-9
153.181
JBJJTCGQCRGNOL-SSDOTTSWSA-N
1268102
C1C=CCC(=C1)C(C(=O)O)N
C8H11NO2
MFCD00137746
r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine
(2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid
Documents