Please login to your online account to display your discounted pricing

2,5-Difluorotoluene, ≥99.5%, Acros Organics


Quantity 25g

Manufacturer: thermo fisher scientific  271510250

 View more versions of this product

Catalog No. AC271510250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.50%
Physical Form Liquid
Formula Weight 128.12
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 05, 224
TSCA TSCA
EINECS Number 207-205-6
Density 1.3600g/mL
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 5-Difluorotoluene
Infrared Spectrum Authentic
Name Note 99.5+%
Molecular Formula C7H6F2
MDL Number MFCD00000347
CAS 452-67-5
CAS Min % 99.5
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Assay Percent Range 99.5+%
Linear Formula CH3C6H3F2

Specifications

Chemical Name or Material 2, 5-Difluorotoluene
Name Note 99.5+%
CAS 452-67-5
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C7H6F2
Linear Formula CH3C6H3F2
Formula Weight 128.12
MDL Number MFCD00000347
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.

SDS & Safety Documents