Please login to your online account to display your discounted pricing

2,5-Difluorotoluene, ≥99.5%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2, 5-Difluorotoluene
Name Note 99.5+%
CAS 452-67-5
CAS Min % 99.5
CAS Max % 100.0
Assay ≥99.50%
Assay Percent Range 99.5+%
Molecular Formula C7H6F2
Linear Formula CH3C6H3F2
Formula Weight 128.12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC271510250

  SDS
thermo fisher scientific
271510250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.50%
Formula Weight 128.12
Physical Form Liquid
Quantity 25g
Color Clear colorless to slightly yellow
Beilstein 05, 224
TSCA TSCA
Density 1.3600g/mL
EINECS Number 207-205-6
CAS Max % 100.0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 99.5
Refractive Index 1.4510 to 1.4530 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
MDL Number MFCD00000347
CAS 452-67-5
Molecular Formula C7H6F2
Chemical Name or Material 2, 5-Difluorotoluene
Name Note 99.5+%
Linear Formula CH3C6H3F2
Assay Percent Range 99.5+%
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Highly flammable liquid and vapor.