Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Difluoropropiophenone, 97%

$33.59 - $557.50

Chemical Identifiers

Chemical Name or Material 2',5'-Difluoropropiophenone
CAS 29112-90-1
Molecular Formula C9H8F2O
Formula Weight 170.16
MDL Number MFCD00015509
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2080103

  SDS
alfa aesar™
B2080103
1g Each for $33.59

AAB2080106

  SDS
alfa aesar™
B2080106
5g Each for $131.09

AAB2080114

  SDS
alfa aesar™
B2080114
25g Each for $557.50
Description & Specifications

Specifications

Formula Weight 170.16
CAS 29112-90-1
Chemical Name or Material 2',5'-Difluoropropiophenone
MDL Number MFCD00015509
Assay 97%
Quantity 1g
Molecular Formula C9H8F2O
Boiling Point 39° to 40°C (0.5mmHg)