Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorophenylhydrazine hydrochloride, 99%

$38.18 - $38.18

Chemical Identifiers

Chemical Name or Material 2,5-Difluorophenylhydrazine hydrochloride
CAS 175135-73-6
Molecular Formula C6H6F2N2·HCl
Formula Weight 180.59
MDL Number MFCD00013385
Beilstein 8640307
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1035903

 
alfa aesar™
L1035903
1g Each for $38.18
Description & Specifications

Specifications

Formula Weight 180.59
Chemical Name or Material 2,5-Difluorophenylhydrazine hydrochloride
Assay 99%
CAS 175135-73-6
Beilstein 8640307
Molecular Formula C6H6F2N2·HCl
MDL Number MFCD00013385
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point ≈211°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.