missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorophenylhydrazine hydrochloride, 99%

Manufacturer:  Alfa Aesar™ L1035903

 View more versions of this product

Catalog No. AAL1035903



Specifications

Specifications

2,5-Difluorophenylhydrazine hydrochloride
1g
C6H7ClF2N2
8640307
LZKLZCSHMDRKJH-UHFFFAOYSA-N
(2,5-difluorophenyl)hydrazine;hydrochloride
24720967
99%
~211°C
175135-73-6
MFCD00013385
2,5-difluorophenylhydrazine hydrochloride, 2,5-difluorophenyl hydrazine hydrochloride, 2,5-difluorophenylhydrazine hcl, 2,5-difluoro-phenyl-hydrazine hydrochloride, hydrazine, 2,5-difluorophenyl-, monohydrochloride, 2,5-difluorphenylhydrazine hydrochloride, 2,5-difluorophenylhy, pubchem1082, acmc-1c9k4, ksc491e8d
C1=CC(=C(C=C1F)NN)F.Cl
180.583
180.59

Chemical Identifiers

175135-73-6
180.583
LZKLZCSHMDRKJH-UHFFFAOYSA-N
24720967
C1=CC(=C(C=C1F)NN)F.Cl
C6H7ClF2N2
MFCD00013385
2,5-difluorophenylhydrazine hydrochloride, 2,5-difluorophenyl hydrazine hydrochloride, 2,5-difluorophenylhydrazine hcl, 2,5-difluoro-phenyl-hydrazine hydrochloride, hydrazine, 2,5-difluorophenyl-, monohydrochloride, 2,5-difluorphenylhydrazine hydrochloride, 2,5-difluorophenylhy, pubchem1082, acmc-1c9k4, ksc491e8d
(2,5-difluorophenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents