Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorophenylhydrazine, 97%

$113.76 - $113.76

Chemical Identifiers

Chemical Name or Material 2,5-Difluorophenylhydrazine
CAS 97108-50-4
Molecular Formula C6H6F2N2
Formula Weight 144.13
MDL Number MFCD00013384
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2125906

 
alfa aesar™
B2125906
5g Each for $113.76
Description & Specifications

Specifications

Formula Weight 144.13
Chemical Name or Material 2,5-Difluorophenylhydrazine
Assay 97%
Molecular Formula C6H6F2N2
MDL Number MFCD00013384
CAS 97108-50-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 73° to 76°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.