missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorophenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 35914-0100

 View more versions of this product

Catalog No. AC359140100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 5-Difluorophenylboronic acid
10g
193353-34-3
100.0
Crystalline Powder
97%
157.91
White to light yellow
Authentic
96.0
Glass bottle
≥96.0%
C6H5BF2O2

Chemical Identifiers

193353-34-3
100.0
C6H5BF2O2
2, 5-Difluorophenylboronic acid
96.0
97%
157.91
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.