missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorophenyl isothiocyanate, 98+%

Manufacturer:  Alfa Aesar™ L1232206

 View more versions of this product

Catalog No. AAL1232206



Specifications

Specifications

2,5-Difluorophenyl isothiocyanate
205°C
206559-57-1
MFCD00041046
6919911
QVIDXDRFMDPVLA-UHFFFAOYSA-N
1,4-difluoro-2-isothiocyanatobenzene
598621
≥98%
92°C (197°F)
5g
C7H3F2NS
UN2922
2,5-difluorophenyl isothiocyanate, 2,5-difluorophenylisothiocyanate, benzene, 1,4-difluoro-2-isothiocyanato, 2,5-difluorobenzenisothiocyanate, pubchem5020, acmc-1cc0g, #, benzene,1,4-difluoro-2-isothiocyanato, 2,5-difluorophenyl isothiocyanate, 1,4-bis fluoranyl-2-isothiocyanato-benzene
C1=CC(=C(C=C1F)N=C=S)F
171.165
171.17

Chemical Identifiers

206559-57-1
171.165
QVIDXDRFMDPVLA-UHFFFAOYSA-N
598621
C1=CC(=C(C=C1F)N=C=S)F
C7H3F2NS
MFCD00041046
2,5-difluorophenyl isothiocyanate, 2,5-difluorophenylisothiocyanate, benzene, 1,4-difluoro-2-isothiocyanato, 2,5-difluorobenzenisothiocyanate, pubchem5020, acmc-1cc0g, #, benzene,1,4-difluoro-2-isothiocyanato, 2,5-difluorophenyl isothiocyanate, 1,4-bis fluoranyl-2-isothiocyanato-benzene
1,4-difluoro-2-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents