Learn More
2,5-Difluorophenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1874414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,5-Difluorophenol | |
36°C to 40°C | |
Phenol-like | |
MFCD00042501 | |
UN2921 | |
phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464 | |
OC1=CC(F)=CC=C1F | |
130.09 | |
130.09 |
2713-31-7 | |
53°C (127°F) | |
C6H4F2O | |
25 g | |
2081076 | |
INXKVYFOWNAVMU-UHFFFAOYSA-N | |
2,5-difluorophenol | |
94952 | |
97% |
Chemical Identifiers
2713-31-7 | |
130.09 | |
INXKVYFOWNAVMU-UHFFFAOYSA-N | |
94952 | |
OC1=CC(F)=CC=C1F |
C6H4F2O | |
MFCD00042501 | |
phenol, 2,5-difluoro, 1,4-difluoro-2-hydroxybenzene, phenol,2,5-difluoro, 2,5-difluorphenol, 2,5-difluorophenol;2,5-difluorophenol;, 2 5-difluorophenol, 2,5-difluoro phenol, 2,5-difluoro-phenol, phenol derivative, 5, pubchem3464 | |
2,5-difluorophenol |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H228
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H228-H302+H312+H332-H314
DOTInformation : Hazard Class: 4.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only