Please login to your online account to display your discounted pricing

2,5-Difluorophenol, 96%, ACROS Organics™

$291.34 - $291.34

Chemical Identifiers

Chemical Name or Material 2, 5-Difluorophenol
Name Note 96%
CAS 2713-31-7
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H4F2O
Linear Formula F2(C6H3)OH
Formula Weight 130.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC241950050

  SDS
thermo fisher scientific
AC241950050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC241950250

  SDS
thermo fisher scientific
241950250
Glass bottle 25g Each for $291.34
Description & Specifications

Specifications

Assay ≥95%
Physical Form Crystalline Mass, Crystals, Needles and/Chunks
Formula Weight 130.1
Quantity 5g
Color White to yellow or beige
TSCA TSCA
Melting Point 35° to 42°C
Beilstein 10, IV, 780
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 2, 5-Difluorophenol
Infrared Spectrum Authentic
Name Note 96%
Assay Percent Range 96%
Molecular Formula C6H4F2O
Linear Formula F2(C6H3)OH
MDL Number MFCD00042501
CAS 2713-31-7
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.