Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluoromandelic acid, 96%

$147.22 - $147.22

Chemical Identifiers

Chemical Name or Material 2,5-Difluoromandelic acid
CAS 207853-61-0
Molecular Formula C8H6F2O3
Formula Weight 188.13
MDL Number MFCD00061297
Synonym alpha-Hydroxy-2,5-difluorophenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1021203

  SDS
alfa aesar™
L1021203
1g Each for $147.22
Description & Specifications

Specifications

Formula Weight 188.13
Chemical Name or Material 2,5-Difluoromandelic acid
CAS 207853-61-0
Molecular Formula C8H6F2O3
Health Hazard 1 Warning
Synonym alpha-Hydroxy-2,5-difluorophenylacetic acid
Melting Point 110° to 113°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 96%
MDL Number MFCD00061297
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.