Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorocinnamic acid, 98%

$45.81 - $152.19

Chemical Identifiers

Chemical Name or Material 2,5-Difluorocinnamic acid
CAS 112898-33-6
Molecular Formula C9H6F2O2
Formula Weight 184.14
MDL Number MFCD00010318
Beilstein 7918450
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1995403

  SDS
alfa aesar™
A1995403
1g Each for $45.81

AAA1995406

  SDS
alfa aesar™
A1995406
5g Each for $152.19
Description & Specifications

Specifications

Formula Weight 184.14
Melting Point 138° to 140°C
Quantity 1g
MDL Number MFCD00010318
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C9H6F2O2
Health Hazard 1 Warning
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 112898-33-6
Chemical Name or Material 2,5-Difluorocinnamic acid
Beilstein 7918450
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.