Learn More
2,5-Difluorobenzylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1653014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,5-Difluorobenzylamine | |
1.222 | |
75°C (167°F) | |
1.495 | |
25 g | |
3539280 | |
2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p | |
NCC1=CC(F)=CC=C1F | |
143.14 | |
143.14 |
85118-06-5 | |
76°C to 78°C (15 mmHg) | |
C7H7F2N | |
MFCD00010143 | |
UN2735 | |
Air Sensitive | |
GDFBHCMFIUBEQT-UHFFFAOYSA-N | |
(2,5-difluorophenyl)methanamine | |
123589 | |
97% |
Chemical Identifiers
85118-06-5 | |
143.14 | |
GDFBHCMFIUBEQT-UHFFFAOYSA-N | |
123589 | |
NCC1=CC(F)=CC=C1F |
C7H7F2N | |
MFCD00010143 | |
2,5-difluorobenzylamine, 2,5-difluorophenyl methanamine, benzenemethanamine, 2,5-difluoro, 2,5-difluorobenzyl amine, 1-2,5-difluorophenyl methanamine, 2, 5-difluorobenzylamine, 2,5-difluorophenyl methylamine, pubchem4401, 2,5-difluoro-benzylamine, acmc-209q4p | |
(2,5-difluorophenyl)methanamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 285-658-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only