Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorobenzyl alcohol, 98%

$76.85 - $260.68

Chemical Identifiers

Chemical Name or Material 2,5-Difluorobenzyl alcohol
CAS 75853-20-2
Molecular Formula C7H6F2O
Formula Weight 144.12
MDL Number MFCD00010627
Beilstein 9043529
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1993206

  SDS
alfa aesar™
A1993206
5g Each for $76.85

AAA1993214

  SDS
alfa aesar™
A1993214
25g Each for $260.68
Description & Specifications

Specifications

Formula Weight 144.12
Chemical Name or Material 2,5-Difluorobenzyl alcohol
Assay 98%
Molecular Formula C7H6F2O
MDL Number MFCD00010627
CAS 75853-20-2
Beilstein 9043529
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 93°C (199°F)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.