Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorobenzotrifluoride, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Difluorobenzotrifluoride
CAS 393-38-4
Formula Weight 182.09
MDL Number MFCD00042494
Beilstein 2049494
Synonym 1,4-Difluoro-2-(trifluoromethyl)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6411103

  SDS
alfa aesar™
H6411103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6411106

  SDS
alfa aesar™
H6411106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 107° to 108°C
Formula Weight 182.09
Refractive Index 1.3940
Density 1.424g/mL
Beilstein 2049494
Chemical Name or Material 2,5-Difluorobenzotrifluoride
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Melting Point −14°C
CAS 393-38-4
Synonym 1,4-Difluoro-2-(trifluoromethyl)benzene
Assay 98%
Color Colorless to pale yellow
Quantity 1g
Storage Note 1 2° to 8°C
MDL Number MFCD00042494
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C