Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorobenzophenone, 97+%

$94.47 - $400.80

Chemical Identifiers

Chemical Name or Material 2,5-Difluorobenzophenone
CAS 85068-36-6
Molecular Formula C13H8F2O
Formula Weight 218.2
MDL Number MFCD00009899
Beilstein 4417185
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2292606

  SDS
alfa aesar™
B2292606
5g Each for $94.47

AAB2292614

  SDS
alfa aesar™
B2292614
25g Each for $400.80
Description & Specifications

Specifications

Formula Weight 218.2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C13H8F2O
Boiling Point 120° to 123°C (0.4mmHg)
Health Hazard 1 Warning
Chemical Name or Material 2,5-Difluorobenzophenone
MDL Number MFCD00009899
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥97%
Flash Point >110°C (230°F)
Quantity 5g
Beilstein 4417185
CAS 85068-36-6
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.