missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorobenzonitrile, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 271550050

 View more versions of this product

Catalog No. AC271550050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 5-Difluorobenzonitrile
Glass Bottle
5g
64248-64-2
100.0
C7H3F2N
MFCD00001777
OJTMHIMQUQOLJV-UHFFFAOYSA-N
2,5-difluorobenzonitrile
123558
Crystalline Low Melting Mass
Authentic
White
99%
98.5
99%
F2C6H3CN
2,5 difluorobenzonitrile, 2,5-difluoro benzonitrile, 2,5-difluoro-benzonitrile, 2,5-difluorobenzenecarbonitrile, 2,5-difluorobenzonitrile, 2,5-diflurobenzonitrile, benzonitrile, 2,5-difluoro, ncr bf ef, pubchem4754
C1=CC(=C(C=C1F)C#N)F
139.105
139.1
≥98.5%

Chemical Identifiers

64248-64-2
139.105
OJTMHIMQUQOLJV-UHFFFAOYSA-N
123558
C1=CC(=C(C=C1F)C#N)F
C7H3F2N
MFCD00001777
2,5 difluorobenzonitrile, 2,5-difluoro benzonitrile, 2,5-difluoro-benzonitrile, 2,5-difluorobenzenecarbonitrile, 2,5-difluorobenzonitrile, 2,5-diflurobenzonitrile, benzonitrile, 2,5-difluoro, ncr bf ef, pubchem4754
2,5-difluorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 264-753-9

TSCA : TSCA

SDS Documents