missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorobenzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC398790050

 View more versions of this product

Catalog No. AC398790050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

212.3
1.58g/mL
100.0
97%
C6H3ClF2O2S
(F)2C6H3SO2Cl
5g
96.0
2, 5-Difluorobenzenesulfonyl chloride
26120-86-5
MFCD00060682
Glass bottle

Chemical Identifiers

26120-86-5
100.0
C6H3ClF2O2S
212.3
2, 5-Difluorobenzenesulfonyl chloride
96.0
97%
(F)2C6H3SO2Cl
MFCD00060682

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger