Please login to your online account to display your discounted pricing

2,5-Difluorobenzenesulfonyl chloride, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC398790010

 View more versions of this product

Catalog No. AC398790010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 212.3
Quantity 1g
Density 1.58g/mL
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 2, 5-Difluorobenzenesulfonyl chloride
Molecular Formula C6H3ClF2O2S
Assay Percent Range 97%
CAS 26120-86-5
MDL Number MFCD00060682
Packaging Glass bottle
Linear Formula (F)2C6H3SO2Cl
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material 2, 5-Difluorobenzenesulfonyl chloride
CAS 26120-86-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H3ClF2O2S
Linear Formula (F)2C6H3SO2Cl
Formula Weight 212.3
MDL Number MFCD00060682
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents