missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluorobenzenesulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

CAS 26120-86-5
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.3
MDL Number MFCD00060682
InChI Key CELLJWUVMKEJDY-UHFFFAOYSA-N
Synonym 2,5-difluoro-benzenesulfonyl chloride, 2,5-difluorobenzene-1-sulfonyl chloride, 2,5-difluorobenzenesulphonyl chloride, 2,5-difluorobenzenesulphonylchloride, acmc-1ch2j, benzenesulfonyl chloride, 2,5-difluoro, ksc201g8t, pubchem5123, zerenex zx009573
PubChem CID 2736911
IUPAC Name 2,5-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)S(=O)(=O)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC398790010
SDS View Documents
Thermo Fisher Scientific
AC398790010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC398790050
SDS View Documents
Thermo Fisher Scientific
AC398790050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

26120-86-5
212.3
CELLJWUVMKEJDY-UHFFFAOYSA-N
2736911
C1=CC(=C(C=C1F)S(=O)(=O)Cl)F
C6H3ClF2O2S
MFCD00060682
2,5-difluoro-benzenesulfonyl chloride, 2,5-difluorobenzene-1-sulfonyl chloride, 2,5-difluorobenzenesulphonyl chloride, 2,5-difluorobenzenesulphonylchloride, acmc-1ch2j, benzenesulfonyl chloride, 2,5-difluoro, ksc201g8t, pubchem5123, zerenex zx009573
2,5-difluorobenzenesulfonyl chloride

Specifications

1.5800g/mL
Glass bottle
221°C
26120-86-5
100.0
C6H3ClF2O2S
MFCD00060682
CELLJWUVMKEJDY-UHFFFAOYSA-N
2,5-difluorobenzenesulfonyl chloride
2736911
97%
110°C
1.58
1g
96.0
96% min. (GC)
(F)2C6H3SO2Cl
2,5-difluoro-benzenesulfonyl chloride, 2,5-difluorobenzene-1-sulfonyl chloride, 2,5-difluorobenzenesulphonyl chloride, 2,5-difluorobenzenesulphonylchloride, acmc-1ch2j, benzenesulfonyl chloride, 2,5-difluoro, ksc201g8t, pubchem5123, zerenex zx009573
C1=CC(=C(C=C1F)S(=O)(=O)Cl)F
212.3
212.3
2, 5-Difluorobenzenesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger