Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluorobenzamide, 98%

$134.41 - $560.51

Chemical Identifiers

Chemical Name or Material 2,5-Difluorobenzamide
CAS 85118-03-2
Molecular Formula C7H5F2NO
Formula Weight 157.12
MDL Number MFCD00015548
Beilstein 1946240
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2097106

  SDS
alfa aesar™
B2097106
5g Each for $134.41

AAB2097114

  SDS
alfa aesar™
B2097114
25g Each for $560.51
Description & Specifications

Specifications

Formula Weight 157.12
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 2,5-Difluorobenzamide
Melting Point 110° to 114°C
Assay 98%
Molecular Formula C7H5F2NO
CAS 85118-03-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
MDL Number MFCD00015548
Beilstein 1946240
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.