Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Difluoroacetophenone, 98%

$43.39 - $141.64

Chemical Identifiers

Chemical Name or Material 2',5'-Difluoroacetophenone
CAS 1979-36-8
Molecular Formula C8H6F2O
Formula Weight 156.13
MDL Number MFCD00009898
Beilstein 2963573
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1391103

  SDS
alfa aesar™
A1391103
1g Each for $43.39

AAA1391106

  SDS
alfa aesar™
A1391106
5g Each for $141.64
Description & Specifications

Specifications

Formula Weight 156.13
Molecular Formula C8H6F2O
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 70°C (158°F)
Quantity 1g
Odor Characteristic
MDL Number MFCD00009898
Chemical Name or Material 2',5'-Difluoroacetophenone
Beilstein 2963573
Boiling Point 190° to 191°C
CAS 1979-36-8
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.