missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluoro-4-nitrobenzoic acid, 97%

Manufacturer:  Alfa Aesar™ H3375706

 View more versions of this product

Catalog No. AAH3375706



Description

Description

Applications
2,5-Difluoro-4-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Specifications

2,5-Difluoro-4-nitrobenzoic acid
145°C to 147°C
116465-48-6
MFCD03425705
GPTNSBLYGCZJQV-UHFFFAOYSA-N
203.101
203.1
97%
Green to White
5g
C7H3F2NO4
2,5-difluoro-4-nitrobenzoic acid, 2,5-difluoro-4-nitrobenzenecarboxylic acid, 2,5-difluoro-4-nitrobenzic acid, 2,5-difluoro-4-nitrobenzoicacid, 2,5-difluoro-4-nitro-benzoic acid, benzoic acid, 2,5-difluoro-4-nitro, 2,5-difluoro-4-nitrobenzenecarboxylicacid, pubchem9789, acmc-2099sr, ksc495m6l
C1=C(C(=CC(=C1F)[N+](=O)[O-])F)C(=O)O
246375
Solid

Chemical Identifiers

116465-48-6
203.101
GPTNSBLYGCZJQV-UHFFFAOYSA-N
246375
C7H3F2NO4
MFCD03425705
2,5-difluoro-4-nitrobenzoic acid, 2,5-difluoro-4-nitrobenzenecarboxylic acid, 2,5-difluoro-4-nitrobenzic acid, 2,5-difluoro-4-nitrobenzoicacid, 2,5-difluoro-4-nitro-benzoic acid, benzoic acid, 2,5-difluoro-4-nitro, 2,5-difluoro-4-nitrobenzenecarboxylicacid, pubchem9789, acmc-2099sr, ksc495m6l
C1=C(C(=CC(=C1F)[N+](=O)[O-])F)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents