missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Difluoro-4-methylbenzoic acid, 95%, Thermo Scientific™

$467.11 - $467.11

Chemical Identifiers

CAS 103877-80-1
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD08063230
InChI Key PWWMQUUDAAWEBK-UHFFFAOYSA-N
Synonym 2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
PubChem CID 13657451
IUPAC Name 2,5-difluoro-4-methylbenzoic acid
SMILES CC1=C(F)C=C(C(O)=O)C(F)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6365206
View Documents Promotion Details
Thermo Scientific Chemicals
H6365206
5 g
Each for $467.11
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

103877-80-1
172.13
PWWMQUUDAAWEBK-UHFFFAOYSA-N
13657451
CC1=C(F)C=C(C(O)=O)C(F)=C1
C8H6F2O2
MFCD08063230
2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
2,5-difluoro-4-methylbenzoic acid

Specifications

103877-80-1
C8H6F2O2
5 g
PWWMQUUDAAWEBK-UHFFFAOYSA-N
2,5-difluoro-4-methylbenzoic acid
13657451
95%
2,5-Difluoro-4-methylbenzoic acid
White
MFCD08063230
2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
CC1=C(F)C=C(C(O)=O)C(F)=C1
172.13
172.13
Solid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'