missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluoro-4-methylbenzoic acid, 95%

Manufacturer:  Alfa Aesar™ H63652MD

 View more versions of this product

Catalog No. AAH63652MD



Specifications

Specifications

2,5-Difluoro-4-methylbenzoic acid
250mg
C8H6F2O2
2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
CC1=C(C=C(C(=C1)F)C(=O)O)F
172.131
172.13
95%
White
103877-80-1
MFCD08063230
PWWMQUUDAAWEBK-UHFFFAOYSA-N
2,5-difluoro-4-methylbenzoic acid
13657451
Solid

Chemical Identifiers

103877-80-1
172.131
PWWMQUUDAAWEBK-UHFFFAOYSA-N
13657451
CC1=C(C=C(C(=C1)F)C(=O)O)F
C8H6F2O2
MFCD08063230
2,5-difluoro-4-methylbenzoicacid, 2,5-difluoro-4-methyl-benzoic acid, benzoic acid,2,5-difluoro-4-methyl, pubchem4944, acmc-1brvm, 4-methyl-2,5-difluorobenzoic acid
2,5-difluoro-4-methylbenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C

SDS Documents