Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Difluoro-4-methoxybenzaldehyde, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Difluoro-4-methoxybenzaldehyde
CAS 879093-08-0
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD06246882
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3191106

  SDS
alfa aesar™
H3191106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.13
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 97° to 100°C
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 98%
Molecular Formula C8H6F2O2
MDL Number MFCD06246882
Quantity 5g
Health Hazard 1 Warning
Chemical Name or Material 2,5-Difluoro-4-methoxybenzaldehyde
CAS 879093-08-0
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.