Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Diethyl-3,4-diphenylcyclopentadienone, 97%

$120.20 - $120.20

Chemical Identifiers

Chemical Name or Material 2,5-Diethyl-3,4-diphenylcyclopentadienone
CAS 51932-77-5
Molecular Formula C21H20O
Formula Weight 288.39g/mol
MDL Number MFCD00013748
Beilstein 2335095
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0022906

 
alfa aesar™
L00229-06
5g Each for $120.20
Description & Specifications

Specifications

Melting Point 99° to 102°C
Quantity 5g
Chemical Name or Material 2,5-Diethyl-3,4-diphenylcyclopentadienone
CAS 51932-77-5
Assay 97%
Molecular Formula C21H20O
Formula Weight 288.39g/mol
MDL Number MFCD00013748
Beilstein 2335095
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C