Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',5'-Diethoxybenzanilide, 97%

$534.91 - $534.91

Chemical Identifiers

Chemical Name or Material 2',5'-Diethoxybenzanilide
CAS 92-22-8
Molecular Formula C17H19NO3
Formula Weight 285.34
MDL Number MFCD00026798
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2022714

  SDS
alfa aesar™
B2022714
25g Each for $534.91
Description & Specifications

Specifications

Formula Weight 285.34
Melting Point 87° to 89°C
Chemical Name or Material 2',5'-Diethoxybenzanilide
Assay 97%
Molecular Formula C17H19NO3
MDL Number MFCD00026798
CAS 92-22-8
Quantity 25g