missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichlorotoluene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 113780250

 View more versions of this product

Catalog No. AC113780250



Description

Description

Specifications

Specifications

2, 5-Dichlorotoluene
79°C
Glass bottle
1.25
Colorless to Yellow
25g
19398-61-9
100.0
C7H6Cl2
MFCD00000609
1,4-dichloro-2-methyl-benzene, 2,5-dichlorotoluene, acmc-209exk, benzene, 1,4-dichloro-2-methyl, benzene,4-dichloro-2-methyl, ksc490m7d, pubchem3700, toluene, 2,5-dichloro, toluene,5-dichloro
KFAKZJUYBOYVKA-UHFFFAOYSA-N
1,4-dichloro-2-methylbenzene
29572
Liquid
1.2500g/mL
Authentic
1.5460 to 1.5480
197.0°C to 200.0°C
4.0°C to 5.0°C
98%
97.5
97.5% min. (GC)
CH3C6H3Cl2
05, 296
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone
CC1=C(C=CC(=C1)Cl)Cl
161.03
161.03
98%

Chemical Identifiers

19398-61-9
161.03
KFAKZJUYBOYVKA-UHFFFAOYSA-N
29572
CC1=C(C=CC(=C1)Cl)Cl
C7H6Cl2
MFCD00000609
1,4-dichloro-2-methyl-benzene, 2,5-dichlorotoluene, acmc-209exk, benzene, 1,4-dichloro-2-methyl, benzene,4-dichloro-2-methyl, ksc490m7d, pubchem3700, toluene, 2,5-dichloro, toluene,5-dichloro
1,4-dichloro-2-methylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 243-032-2

TSCA : TSCA

SDS Documents