Please login to your online account to display your discounted pricing

2,5-Dichlorotoluene, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  113780050

 View more versions of this product

Catalog No. AC113780050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 161.03
Quantity 5g
Color Clear colorless to light yellow
Beilstein 05, 296
TSCA TSCA
EINECS Number 243-032-2
Density 1.2500g/mL
CAS Max % 100.0
Refractive Index 1.5460 to 1.5480 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00000609
Name Note 98%
Linear Formula CH3C6H3Cl2
CAS 19398-61-9
Chemical Name or Material 2, 5-Dichlorotoluene
Molecular Formula C7H6Cl2

Specifications

Chemical Name or Material 2, 5-Dichlorotoluene
Name Note 98%
CAS 19398-61-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6Cl2
Linear Formula CH3C6H3Cl2
Formula Weight 161.03
MDL Number MFCD00000609
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents