missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichlorotoluene, 98%, ACROS Organics™

$125.51 - $125.51

Chemical Identifiers

CAS 19398-61-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H6Cl2
Linear Formula CH3C6H3Cl2
Formula Weight 161.03
MDL Number MFCD00000609
Chemical Name or Material 2, 5-Dichlorotoluene, 98%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC113780050 SDS Thermo Fisher Scientific
113780050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC113780250 SDS Thermo Fisher Scientific
113780250
Glass bottle 25g Each for $125.51
Specifications Description & Specifications

Chemical Identifiers

19398-61-9
100.0
C7H6Cl2
161.03
2, 5-Dichlorotoluene, 98%
97.5
98%
CH3C6H3Cl2
MFCD00000609
≥97.5%

Specifications

Liquid
5g
05, 296
100.0
97.5
Authentic
MFCD00000609
19398-61-9
2, 5-Dichlorotoluene, 98%
161.03
Clear colorless to light yellow
1.2500g/mL
1.5460 to 1.5480 (20°C, 589nm)
Glass bottle
98%
CH3C6H3Cl2
C7H6Cl2
≥97.5%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 243-032-2

TSCA : TSCA