Learn More
2,5-Dichlorothiophenol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1143414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,5-Dichlorothiophenol | |
22°C to 25°C | |
>110°C (230°F) | |
C6H4Cl2S | |
MFCD00004831 | |
2042866 | |
2,5-dichlorothiophenol, 2,5-dichlorobenzene-1-thiol, benzenethiol, 2,5-dichloro, 2,5-dichloro thiophenol, thiophenol, 2,5-dichloro, 2,5-dichlorophenylmercaptan, pubchem6801, thiophenol,5-dichloro, 2,5-dichlorobenzenthiol, 2,5-dichloro-thiophenol | |
C1=CC(=C(C=C1Cl)S)Cl | |
179.058 | |
179.07 |
5858-18-4 | |
112°C to 116°C (50 mmHg) | |
Unpleasant | |
1.619 | |
25 g | |
Air Sensitive | |
QIULLHZMZMGGFH-UHFFFAOYSA-N | |
2,5-dichlorobenzenethiol | |
79965 | |
98% |
Chemical Identifiers
5858-18-4 | |
179.058 | |
QIULLHZMZMGGFH-UHFFFAOYSA-N | |
79965 | |
C1=CC(=C(C=C1Cl)S)Cl |
C6H4Cl2S | |
MFCD00004831 | |
2,5-dichlorothiophenol, 2,5-dichlorobenzene-1-thiol, benzenethiol, 2,5-dichloro, 2,5-dichloro thiophenol, thiophenol, 2,5-dichloro, 2,5-dichlorophenylmercaptan, pubchem6801, thiophenol,5-dichloro, 2,5-dichlorobenzenthiol, 2,5-dichloro-thiophenol | |
2,5-dichlorobenzenethiol |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 227-486-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only