Please login to your online account to display your discounted pricing

2,5-Dichlorothiophene 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 5-Dichlorothiophene
Name Note 98%
CAS 3172-52-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4 H2 Cl2 S
Formula Weight 153.03
MDL Number MFCD00005423
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC113770500

  SDS
thermo fisher scientific
113770500
Glass bottle 50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.03
Physical Form Liquid
Assay ≥97.5%
Quantity 50mL
Color Clear colorless to slightly yellow
EINECS Number 221-638-8
TSCA TSCA
Density 1.4400g/mL
Beilstein 17, 33
CAS Max % 100.0
Chemical Name or Material 2, 5-Dichlorothiophene
Storage Information this product darkens on storage
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4 H2 Cl2 S
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5610 to 1.5630 (20°C, 589nm)
MDL Number MFCD00005423
CAS 3172-52-9
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.