Please login to your online account to display your discounted pricing

2,5-Dichlorothiophene 98%, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  113770500

 View more versions of this product

Catalog No. AC113770500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 153.03
Assay ≥97.5%
Physical Form Liquid
Quantity 50mL
Color Clear colorless to slightly yellow
EINECS Number 221-638-8
TSCA TSCA
Density 1.4400g/mL
Beilstein 17, 33
CAS Max % 100.0
Packaging Glass bottle
Storage Information this product darkens on storage
CAS Min % 97.5
Refractive Index 1.5610 to 1.5630 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Suspected of causing cancer.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
Name Note 98%
Chemical Name or Material 2, 5-Dichlorothiophene
Molecular Formula C4 H2 Cl2 S
MDL Number MFCD00005423
CAS 3172-52-9

Specifications

Chemical Name or Material 2, 5-Dichlorothiophene
Name Note 98%
CAS 3172-52-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4 H2 Cl2 S
Formula Weight 153.03
MDL Number MFCD00005423
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing cancer.
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Flammable liquid and vapor.

SDS & Safety Documents