Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorothiophene, 98%

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorothiophene
CAS 3172-52-9
Molecular Formula C4H2Cl2S
Formula Weight 153.03
MDL Number MFCD00005423
UN Number UN2929
Beilstein 109887
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1383914

  SDS
alfa aesar™
A1383914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1383922

  SDS
alfa aesar™
A1383922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1383936

  SDS
alfa aesar™
A1383936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.03
Chemical Name or Material 2,5-Dichlorothiophene
TSCA TSCA
Assay 98%
Molecular Formula C4H2Cl2S
MDL Number MFCD00005423
CAS 3172-52-9
UN Number UN2929
Beilstein 109887
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H226-H341
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
Flash Point 59°C (138°F)
Odor Pungent
Boiling Point 160° to 162°C
Melting Point −41°C
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H226-H341
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Suspected of causing genetic defects.

2° to 8°C