Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorothiophene-3-carboxylic acid, 98+%

$84.99 - $212.46

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorothiophene-3-carboxylic acid
CAS 36157-41-2
Molecular Formula C5H2Cl2O2S
Formula Weight 197.04
MDL Number MFCD00082908
Beilstein 127865
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1910104

  SDS
alfa aesar™
L1910104
2g Each for $212.46

AAL19101ME

  SDS
alfa aesar™
L19101ME
500mg Each for $84.99
Description & Specifications

Specifications

Formula Weight 197.04
CAS 36157-41-2
Molecular Formula C5H2Cl2O2S
Assay ≥98%
MDL Number MFCD00082908
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 147° to 149°C
Quantity 2g
Beilstein 127865
Chemical Name or Material 2,5-Dichlorothiophene-3-carboxylic acid
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.