missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichlorothiophene-3-carbonyl chloride, 95%, Maybridge™

Manufacturer:  Fisher Scientific Company CD09458EA

 View more versions of this product

Catalog No. CD09458EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2, 5-Dichlorothiophene-3-carbonyl chloride
10g
94.5
95%
MFCD00041441
YGDFSMANOSKQRU-UHFFFAOYSA-N
2,5-dichlorothiophene-3-carbonyl chloride
2777084
95%
Amber Glass Bottle
57248-14-3
100.0
C5HCl3OS
2,5-bis chloranyl thiophene-3-carbonyl chloride, 2,5-dichloro-3-thenoyl chloride, 2,5-dichloro-3-thiophenecarbonyl chloride, 2,5-dichloro-3-thiophenecarbonylchloride, 2,5-dichloro-thiophene-3-carbonyl chloride, 2,5-dichlorothiophene-3-carbonylchloride, 3-thiophenecarbonylchloride, 2,5-dichloro, pubchem15832
C1=C(SC(=C1C(=O)Cl)Cl)Cl
215.472
215.49

Chemical Identifiers

57248-14-3
215.472
YGDFSMANOSKQRU-UHFFFAOYSA-N
2777084
C1=C(SC(=C1C(=O)Cl)Cl)Cl
C5HCl3OS
MFCD00041441
2,5-bis chloranyl thiophene-3-carbonyl chloride, 2,5-dichloro-3-thenoyl chloride, 2,5-dichloro-3-thiophenecarbonyl chloride, 2,5-dichloro-3-thiophenecarbonylchloride, 2,5-dichloro-thiophene-3-carbonyl chloride, 2,5-dichlorothiophene-3-carbonylchloride, 3-thiophenecarbonylchloride, 2,5-dichloro, pubchem15832
2,5-dichlorothiophene-3-carbonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents