missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloropyridine, 98%

Manufacturer:  Alfa Aesar™ A1544214

 View more versions of this product

Catalog No. AAA1544214



Description

Description

Applications
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.

Solubility
Insoluble in water.

Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
Specifications

Specifications

2,5-Dichloropyridine
Unpleasant
59°C to 62°C
16110-09-1
MFCD00006239
108886
GCTFDMFLLBCLPF-UHFFFAOYSA-N
2,5-dichloropyridine
27685
98%
112°C (233°F)
190°C to 191°C
25g
C5H3Cl2N
UN2811
pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
C1=CC(=NC=C1Cl)Cl
147.986
147.99

Chemical Identifiers

16110-09-1
147.986
GCTFDMFLLBCLPF-UHFFFAOYSA-N
27685
C1=CC(=NC=C1Cl)Cl
C5H3Cl2N
MFCD00006239
pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine
2,5-dichloropyridine
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309+P311-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : US8225000

Documents