Learn More
2,5-Dichloropyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1544218
Description
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.
Solubility
Insoluble in water.
Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
Specifications
2,5-Dichloropyridine | |
59°C to 62°C | |
112°C (233°F) | |
C5H3Cl2N | |
50 g | |
108886 | |
Insoluble in water. | |
ClC1=CC=C(Cl)N=C1 | |
147.99 | |
147.99 |
16110-09-1 | |
190°C to 191°C | |
Unpleasant | |
MFCD00006239 | |
UN2811 | |
pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine | |
GCTFDMFLLBCLPF-UHFFFAOYSA-N | |
2,5-dichloropyridine | |
27685 | |
98% |
Chemical Identifiers
16110-09-1 | |
147.99 | |
GCTFDMFLLBCLPF-UHFFFAOYSA-N | |
27685 | |
ClC1=CC=C(Cl)N=C1 |
C5H3Cl2N | |
MFCD00006239 | |
pyridine, 2,5-dichloro, 2,5-dichloro-pyridine, 2,5-dichloro pyridine, ccris 1718, 2,5-dichlorpyridin, 2,5-dichloropyridin, 2,5-dichioropyridine, 2.5-dichloropyridine, pubchem1199, 2,5 dichloropyridine | |
2,5-dichloropyridine |
Safety and Handling
GHS H Statement
H311-H302-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 240-278-2
RTECSNumber : US8225000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only