Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichloropyridine-4-boronic acid pinacol ester, 95%

$215.47 - $488.19

Chemical Identifiers

Chemical Name or Material 2,5-Dichloropyridine-4-boronic acid pinacol ester
CAS 1073353-98-6
Molecular Formula C11H14BCl2NO2
Formula Weight 273.96
MDL Number MFCD06798256
Synonym 2,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2769903

  SDS
alfa aesar™
H2769903
1g Each for $488.19

AAH27699MD

  SDS
alfa aesar™
H27699MD
250mg Each for $215.47
Description & Specifications

Specifications

Formula Weight 273.96
Synonym 2,5-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula C11H14BCl2NO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2,5-Dichloropyridine-4-boronic acid pinacol ester
MDL Number MFCD06798256
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 83° to 85°C
CAS 1073353-98-6
Health Hazard 1 Warning
Quantity 1g
Assay 95%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.