Please login to your online account to display your discounted pricing

2,5-Dichlorophenylhydrazine, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  165470050

 View more versions of this product

Catalog No. AC165470050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (GC)
Formula Weight 177.03
Physical Form Powder
Quantity 5g
Color White to yellow
TSCA TSCA
EINECS Number 206-163-6
RTECS Number MV2100000
Melting Point 99° to 104°C
Beilstein 15, 431
CAS Max % 100.0
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 305-15-7
Name Note 98%
Molecular Formula C6H6Cl2N2
Linear Formula Cl2C6H3NHNH2
Assay Percent Range 98%
MDL Number MFCD00007576
Chemical Name or Material 2, 5-Dichlorophenylhydrazine

Specifications

Chemical Name or Material 2, 5-Dichlorophenylhydrazine
Name Note 98%
CAS 305-15-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H6Cl2N2
Linear Formula Cl2C6H3NHNH2
Formula Weight 177.03
MDL Number MFCD00007576
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.