Please login to your online account to display your discounted pricing

2,5-Dichlorophenylhydrazine, 98%, ACROS Organics™

$50.85 - $50.85

Chemical Identifiers

Chemical Name or Material 2, 5-Dichlorophenylhydrazine
Name Note 98%
CAS 305-15-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H6Cl2N2
Linear Formula Cl2C6H3NHNH2
Formula Weight 177.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC165470250

  SDS
thermo fisher scientific
165470250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC165470050

  SDS
thermo fisher scientific
165470050
Glass bottle 5g Each for $50.85
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5% (GC)
Formula Weight 177.03
Quantity 25g
Color White to yellow
Beilstein 15, 431
EINECS Number 206-163-6
TSCA TSCA
RTECS Number MV2100000
Melting Point 99° to 104°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2, 5-Dichlorophenylhydrazine
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H6Cl2N2
Linear Formula Cl2C6H3NHNH2
MDL Number MFCD00007576
CAS 305-15-7
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.