Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorophenylhydrazine, 98%

$49.27 - $153.69

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorophenylhydrazine
CAS 305-15-7
Molecular Formula C6H6Cl2N2
Formula Weight 177.03
MDL Number MFCD00007576
Beilstein 743382
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0670806

  SDS
alfa aesar™
L0670806
5g Each for $49.27

AAL0670814

  SDS
alfa aesar™
L0670814
25g Each for $153.69
Description & Specifications

Specifications

Formula Weight 177.03
Health Hazard 1 Warning
Chemical Name or Material 2,5-Dichlorophenylhydrazine
TSCA TSCA
Assay 98%
Molecular Formula C6H6Cl2N2
MDL Number MFCD00007576
CAS 305-15-7
Beilstein 743382
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
RTECS Number MV2100000
Quantity 5g
Melting Point 102° to 105°C
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C