missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichlorophenylboronic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC393840050

 View more versions of this product

Catalog No. AC393840050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2, 5-Dichlorophenylboronic acid
5g
Authentic
100.0
Powder
97%
190.82
White to off-white
135145-90-3
96.0
Glass bottle
≥96.0%
C6H5BCl2O2
MFCD01863182

Chemical Identifiers

135145-90-3
100.0
C6H5BCl2O2
MFCD01863182
≥96.0%
96.0
97%
190.82
2, 5-Dichlorophenylboronic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.