missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorophenyl isocyanate, 97%

Manufacturer:  Alfa Aesar™ L0973506

 View more versions of this product

Catalog No. AAL0973506



Specifications

Specifications

2,5-Dichlorophenyl isocyanate
Irritating
28°C to 31°C
5392-82-5
MFCD00002002
388960
PEQMJVGRHNZPAM-UHFFFAOYSA-N
1,4-dichloro-2-isocyanatobenzene
79360
97%
>110°C (230°F)
120°C to 122°C (20mmHg)
5g
C7H3Cl2NO
UN2250
2,5-dichlorophenyl isocyanate, benzene, 1,4-dichloro-2-isocyanato, 2,5-dichlorophenylisocyanate, isocyanic acid, 2,5-dichlorophenyl ester, isocyanic acid 2,5-dichlorophenyl ester, acmc-209laz, 2,5-dichlorobenzenisocyanate, benzene,4-dichloro-2-isocyanato, #, isocyanic acid,5-dichlorophenyl ester
C1=CC(=C(C=C1Cl)N=C=O)Cl
188.007
188.01

Chemical Identifiers

5392-82-5
188.007
PEQMJVGRHNZPAM-UHFFFAOYSA-N
79360
C1=CC(=C(C=C1Cl)N=C=O)Cl
C7H3Cl2NO
MFCD00002002
2,5-dichlorophenyl isocyanate, benzene, 1,4-dichloro-2-isocyanato, 2,5-dichlorophenylisocyanate, isocyanic acid, 2,5-dichlorophenyl ester, isocyanic acid 2,5-dichlorophenyl ester, acmc-209laz, 2,5-dichlorobenzenisocyanate, benzene,4-dichloro-2-isocyanato, #, isocyanic acid,5-dichlorophenyl ester
1,4-dichloro-2-isocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents