Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,5-Dichlorophenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2,5-Dichlorophenyl isocyanate
CAS 5392-82-5
Molecular Formula C7H3Cl2NO
Formula Weight 188.01
MDL Number MFCD00002002
UN Number UN2250
Beilstein 388960
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0973506

  SDS
alfa aesar™
L0973506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0973514

  SDS
alfa aesar™
L0973514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.01
Chemical Name or Material 2,5-Dichlorophenyl isocyanate
Molecular Formula C7H3Cl2NO
CAS 5392-82-5
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
TSCA TSCA
Odor Irritating
Assay 97%
MDL Number MFCD00002002
UN Number UN2250
Boiling Point 120° to 122°C (20mmHg)
Beilstein 388960
Melting Point 28° to 31°C
Flash Point >110°C (230°F)
Health Hazard 3 GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C